Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tide Quencher™ 3 maleimide
Tide Quencher™ 3 maleimide, Tide Quencher™ fluorescent probe
Tide Quencher™ 3 maleimide, Tide Quencher™ fluorescent probe
Irritant
Tide Quencher™ 3 maleimide, Tide Quencher™ fluorescent probe
Catalog # AATB2226
Supplier:
AAT Bioquest
CAS Number:
Expand image
Irritant
Tide Quencher™ 3 maleimide, Tide Quencher™ fluorescent probe
Catalog # AATB2226
Supplier:
AAT Bioquest
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
5 mg
Seq:
2210
Specifications
About this item
Product Details
:
:
575.64 g/mol
ADR:
5,II
:
GHS07
Product Details