Order Entry
Switzerland
ContactUsLinkComponent
2,4,6-Trimethylphenylmagnesium bromide ≥95%
2,4,6-Trimethylphenylmagnesium bromide ≥95%
Catalog # AATB22101
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
2,4,6-Trimethylphenylmagnesium bromide ≥95%
Catalog # AATB22101
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :223.396 g/mol
CAS Number:2633-66-1
Formula:C₉H₁₁BrMg
MDL Number:MFCD00009663