Order Entry
Switzerland
ContactUsLinkComponent
3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)] ≥95%
3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)] ≥95%
Catalog # AATB22076
Supplier:  AAT Bioquest
CAS Number:  
undefined
3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)] ≥95%
Catalog # AATB22076
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :546.53 g/mol
CAS Number:53213-94-8
Formula:C₂₅H₂₇IN₂S₂
MDL Number:MFCD00075033