Order Entry
Switzerland
ContactUsLinkComponent
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindotricarbocyanine iodide (DiR) ≥95%
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindotricarbocyanine iodide (DiR) ≥95%
Catalog # AATB22070
Supplier:  AAT Bioquest
CAS Number:  
undefined
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindotricarbocyanine iodide (DiR) ≥95%
Catalog # AATB22070
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :1013.39 g/mol
CAS Number:100068-60-8
Formula:C₆₃H₁₀₁N₂I
MDL Number:MFCD03427788