Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
DiIC1(5) iodide
DiIC1(5) iodide ≥95%
DiIC1(5) iodide ≥95%
DiIC1(5) iodide ≥95%
Catalog # AATB22056
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
DiIC1(5) iodide ≥95%
Catalog # AATB22056
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pk:
25 mg
Seq:
2315
Specifications
About this item
Product Details
:
:
510.5 g/mol
CAS Number:
36536-22-8
Formula:
C₂₇H₃₁IN₂
Product Details