Order Entry
Switzerland
ContactUsLinkComponent
5(6)-Carboxy-2′,7′- dichlorofluorescein diacetate mixture of isomers ≥95%
5(6)-Carboxy-2′,7′- dichlorofluorescein diacetate mixture of isomers ≥95%
Catalog # AATB22025
Supplier:  AAT Bioquest
CAS Number:  
undefined
5(6)-Carboxy-2′,7′- dichlorofluorescein diacetate mixture of isomers ≥95%
Catalog # AATB22025
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :529.28 g/mol
CAS Number:127770-45-0
Formula:C₂₅H₁₄Cl₂O₉
MDL Number:MFCD08459585