Order Entry
Switzerland
ContactUsLinkComponent
BCECF AM ester, CytoFix™ Optimized for long term cellular pH tracking
Irritant
BCECF AM ester, CytoFix™ Optimized for long term cellular pH tracking
Catalog # AATB21200
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
BCECF AM ester, CytoFix™ Optimized for long term cellular pH tracking
Catalog # AATB21200
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :808.69 g/mol
ADR:5,II
 :GHS07
CAS Number:117464-70-7
Formula:C₄₂H₄₀O₂₁
MDL Number:MFCD00036969