Order Entry
Switzerland
ContactUsLinkComponent
 
Irritant
Cal-520L®/Cy5-Dextran conjugate, M.W. 10,000, RatioWorks™
Catalog # AATB20603
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
Cal-520L®/Cy5-Dextran conjugate, M.W. 10,000, RatioWorks™
Catalog # AATB20603
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :12000 g/mol
ADR:5,II
 :GHS07