Order Entry
Switzerland
ContactUsLinkComponent
Furaptra (Mag-Fura-2) tetrapotassium salt magnesium indicator
Irritant
Furaptra (Mag-Fura-2) tetrapotassium salt magnesium indicator
Catalog # AATB20383
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
Furaptra (Mag-Fura-2) tetrapotassium salt magnesium indicator
Catalog # AATB20383
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 µG
  • Seq:
    1418

Specifications

About this item

 :
ADR:5,II
 :587 g/mol
 :GHS07
CAS Number:132319-57-4
Formula:C₁₈H₁₀K₄N₂O₁₁