Order Entry
Switzerland
ContactUsLinkComponent
 
Probenecid 25 mM in aqueous solution stabilised, ReadiUse™
Catalog # AATB20062
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Probenecid 25 mM in aqueous solution stabilised, ReadiUse™
Catalog # AATB20062
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :285.4 g/mol
CAS Number:57-66-9
Formula:C₁₃H₁₉NO₄S
MDL Number:MFCD00038402