Order Entry
Switzerland
ContactUsLinkComponent
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) 10 mM in water ≥95%
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) 10 mM in water ≥95%
Catalog # AATB17507
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) 10 mM in water ≥95%
Catalog # AATB17507
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    2 mL
  • Seq:
    2145

Specifications

About this item

 :
 :350.25 g/mol
CAS Number:28718-90-3
Formula:C₁₆H₁₅N₅·2HCl
MDL Number:MFCD00012681