Order Entry
Switzerland
ContactUsLinkComponent
cAMP-NHS ester
cAMP-NHS ester
Catalog # AATB17102
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
cAMP-NHS ester
Catalog # AATB17102
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :684.64 g/mol
ADR:5,II
 :GHS07