
Aminopropargyl ddCTP for Sanger sequencing
Catalog # AATB17070
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:10 µmol
- Seq:9105
Specifications
About this item
:
:592.15 g/mol
ADR:5,II
:GHS07
CAS Number:114748-56-0