
DAF-2 for nitric oxide detection
Catalog # AATB16290
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 mg
- Seq:2070
Specifications
About this item
:
:364.34 g/mol
ADR:5,II
:GHS07
CAS Number:205391-01-1
Formula:C₂₀H₁₆N₂O₅
MDL Number:MFCD01866182