Order Entry
Switzerland
ContactUsLinkComponent
Cyanine 7 monosuccinimidyl ester ≥90% [equivalent to Cy7® NHS ester]
Cyanine 7 monosuccinimidyl ester ≥90% [equivalent to Cy7® NHS ester]
Catalog # AATB161
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Cyanine 7 monosuccinimidyl ester ≥90% [equivalent to Cy7® NHS ester]
Catalog # AATB161
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :779.92 g/mol
CAS Number:477908-53-5
Formula:C₃₉H₄₅N₃O₁₀S₂
MDL Number:MFCD28968779