Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cyanine 3 amine
Cyanine 3 amine ≥95% [equivalent to Cy3® amine]
Cyanine 3 amine ≥95% [equivalent to Cy3® amine]
Cyanine 3 amine ≥95% [equivalent to Cy3® amine]
Catalog # AATB145
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
Cyanine 3 amine ≥95% [equivalent to Cy3® amine]
Catalog # AATB145
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
900.9 g/mol
Product Details