Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Auftragserfassung
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Heim
TF3-DEVD-FMK
TF3-DEVD-FMK
TF3-DEVD-FMK
Irritant
TF3-DEVD-FMK
Catalog # AATB13474
Supplier:
AAT Bioquest
CAS Number:
Expand image
Irritant
TF3-DEVD-FMK
Catalog # AATB13474
Supplier:
AAT Bioquest
CAS Number:
Specifications
Packung:
Plastic bottle
VE:
25 Tests
Seq:
0770
Specifications
About this item
Product Details
:
ADR:
5,II
:
GHS07
Product Details