Order Entry
Switzerland
ContactUsLinkComponent
L-Leucine(4-methyl-7-coumarinylamide) hydrochloride ≥95%
Irritant
L-Leucine(4-methyl-7-coumarinylamide) hydrochloride ≥95%
Catalog # AATB13445
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
L-Leucine(4-methyl-7-coumarinylamide) hydrochloride ≥95%
Catalog # AATB13445
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :324.805 g/mol
ADR:5,II
 :GHS07
CAS Number:62480-44-8
Formula:C₁₆H₂₀N₂O₃·HCl
MDL Number:MFCD00043262