Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-FITC cadaverine
5-FITC cadaverine ≥95%
5-FITC cadaverine ≥95%
Catalog # AATB129
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
5-FITC cadaverine ≥95%
Catalog # AATB129
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pk:
100 mg
Seq:
2440
Specifications
About this item
Product Details
:
:
491.56 g/mol
Formula:
C₂₆H₂₅N₃O₅S
Product Details