Order Entry
Switzerland
ContactUsLinkComponent
Luciferase for luminescent imaging
Irritant
Luciferase for luminescent imaging
Catalog # AATB12500
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
Luciferase for luminescent imaging
Catalog # AATB12500
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
 :60 kDa
ADR:5,II
 :GHS07
CAS Number:61970-00-1