Order Entry
Switzerland
ContactUsLinkComponent
Native Glucose-UDP (from Eukaryotic cells), Fluorescein
Native Glucose-UDP (from Eukaryotic cells), Fluorescein
Catalog # AATB11705
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Native Glucose-UDP (from Eukaryotic cells), Fluorescein
Catalog # AATB11705
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    100 µG
  • Conjugation:
    Fluorescein
  • Protein/peptide type:
    Native
  • Source:
    Eukaryotic cells
  • Protein synonyms:
    Uridine-5'-diphospho-1-alpha-D-glucose

Specifications

About this item

Uridine-5'-diphospho-1-alpha-D-glucose (UDP-Glc) is a key intermediate in carbohydrate metabolism.

  • Metabolic precursor
  • Well studied substrate
  • Multiple applications
Caution: For research use only.