Native cAMP (from Eukaryotic cells), Biotin
Référence AATB17105
Fournisseur: AAT BIOQUEST
Numéro CAS:
Spécifications
- Pack type:Vial
- Pk:100 µG
- Conjugation:Biotin
- Protein/peptide type:Native
- Source:Eukaryotic cells
- Protein synonyms:Cyclic adenosine monophosphate, Cyclic AMP
- Protein/peptide name:cAMP
Spécifications
À propos de cet article
Cyclic adenosine monophosphate (cAMP) is a second messenger used for intracellular signal induction.
- Essential component of intracellular signaling
Attention: For research use only.