Native Armoracia rusticana HRP (from Armoracia rusticana), Estradiol
Catalog # AATB50550
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Vial
- Pk:1 mg
- Conjugation:Estradiol
- Protein/peptide type:Native
- Source:Armoracia rusticana
- Species:Armoracia rusticana
- Protein synonyms:Horseradish Peroxidase
- Protein/peptide name:HRP
Specifications
About this item
Estradiol is an estrogen steroid hormone and the major female sex hormone.
- Assay development
- High sensitivity
Caution: For research use only.