Order Entry
Switzerland
ContactUsLinkComponent
Native Human Wheat Germ Agglutinin (from Eukaryotic cells), iFluor® 594
Native Human Wheat Germ Agglutinin (from Eukaryotic cells), iFluor® 594
Catalog # AATB25550
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Native Human Wheat Germ Agglutinin (from Eukaryotic cells), iFluor® 594
Catalog # AATB25550
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    1 mg
  • Conjugation:
    iFluor® 594
  • Protein/peptide type:
    Native
  • Source:
    Eukaryotic cells
  • Species:
    Human
  • Protein synonyms:
    WGA
  • Protein/peptide name:
    Wheat Germ Agglutinin

Specifications

About this item

Wheat germ agglutinin (WGA) is a lectin that binds to N-acetyl-D-glucosamine and sialic acid.

  • Glycoconjugate binding
  • Very bright signal
  • Multiple applications
Caution: For research use only.