ContactUsLinkComponent
ddTTP [2',3'-Dideoxythymidine-5'-triphosphate] *10 mM in ddH₂O* (1 µmole)
ddTTP [2',3'-Dideoxythymidine-5'-triphosphate] *10 mM in ddH₂O* (1 µmole)
Catalog # AATB17208
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
ddTTP [2',3'-Dideoxythymidine-5'-triphosphate] *10 mM in ddH₂O* (1 µmole)
Catalog # AATB17208
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    1 µmol
  • Description:
    ddTTP [2',3'-dideoxythymidine-5'-triphosphate] *10 mm in ddH₂O*
Specifications
About this item
Sanger sequencing, also known as the chain termination method, is a technique for DNA sequencing based upon the selective incorporation of chain-terminating dideoxynucleotides (ddNTPs) by DNA polymerase.
  • In-house quality control
  • Useful for sanger sequencing applications
Caution: For research use only.