Order Entry
Switzerland
ContactUsLinkComponent
Texas Red® Hydrazide, Single Isomer
Texas Red® Hydrazide, Single Isomer
Catalog # AATB481
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Texas Red® Hydrazide, Single Isomer
Catalog # AATB481
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Description:
    Texas Red® hydrazide
  • Purity:
    95%

Specifications

About this item

Texas Red® hydrazide reacts with carbonyl compounds such as aldehyde, ketones and acids to give bright red fluorescent conjugates.

  • Identical spectra to Texas Red
  • High Stability
Caution: For research use only.