ContactUsLinkComponent
2-Aminoethoxypropargyl ddGTP (1 µmole)
2-Aminoethoxypropargyl ddGTP (1 µmole)
Catalog # AATB17086
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
2-Aminoethoxypropargyl ddGTP (1 µmole)
Catalog # AATB17086
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    1 µmol
  • Description:
    2-aminoethoxypropargyl ddGTP
Specifications
About this item
Sanger method is one of the most reliable and earliest DNA sequencing methods.
  • In-house quality control
  • Useful for sanger sequencing applications
Caution: For research use only.