ContactUsLinkComponent
Tetracysteine REASH Reagent *2 mM DMSO Solution*
Tetracysteine REASH Reagent *2 mM DMSO Solution*
Catalog # AATB22332
Supplier:  AAT Bioquest
CAS Number:  
undefined
Tetracysteine REASH Reagent *2 mM DMSO Solution*
Catalog # AATB22332
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Description:
    Tetracysteine REASH reagent *2 mm DMSO solution*
  • Tests per kit:
    100 Tests
Specifications
About this item
REASH is a resorufin derivative, modified to contain two arsenic atoms at a set distance from each other.
  • Resorufin derivative
  • High affinity for thiols
  • Multiple applications
Caution: For research use only.