ContactUsLinkComponent
ddCTP [2',3'-Dideoxycytidine-5'-triphosphate] *10 mM in ddH₂O* (1 µmole)
ddCTP [2',3'-Dideoxycytidine-5'-triphosphate] *10 mM in ddH₂O* (1 µmole)
Référence AATB17207
Fournisseur:  AAT BIOQUEST
Numéro CAS:  
undefined
ddCTP [2',3'-Dideoxycytidine-5'-triphosphate] *10 mM in ddH₂O* (1 µmole)
Référence AATB17207
Fournisseur:  AAT BIOQUEST
Numéro CAS:  
Spécifications
  • Pk:
    1 µmol
  • Description:
    ddCTP [2',3'-dideoxycytidine-5'-triphosphate] *10 mm in ddH₂O*
Spécifications
À propos de cet article
Sanger sequencing, also known as the chain termination method, is a technique for DNA sequencing based upon the selective incorporation of chain-terminating dideoxynucleotides (ddNTPs) by DNA polymerase.
  • In-house quality control
  • Useful for sanger sequencing applications
Attention: For research use only.