ReadiLink™ Rapid iFluor® 790 Antibody Labeling Kit "Microscale Optimized for Labeling 50 µg Antibody Per Reaction" (2 Labelings)
Catalog # AATB1265
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:2 Tests
- Description:ReadiLink™ rapid iFluor™ 790 antibody labelling kit 'Microscale optimized for labelling 50 µg antibody per reaction'
Specifications
About this item
AAT Bioquest's iFluor® dyes are developed for labelling proteins, in particular, antibodies.
- Contains all essential components
- Minimal hands on time
- Ready to use
- Robust and simple antibody labelling protocol
- No column purification required
Caution: For research use only.