Buccutite™ Rapid Protein Crosslinking Kit *Microscale Optimized for Crosslinking 100 ug Antibody Per Reaction* (2 Labelings)
AATB1315
:
- Pk:2 Tests
- Description:Buccutite™ rapid protein crosslinking kit 'Microscale optimized for crosslinking 100 µg antibody per reaction'
Protein-protein conjugations are commonly performed with a bifunctional linker such as SMCC.
- No catalyst required
- Neutral crosslinking
- Robust and simple protocol
: For research use only.