Order Entry
Switzerland
ContactUsLinkComponent
ReadiLink™ xtra Rapid iFluor® 488 Antibody Labeling Kit *BSA-Compatible* (2 Labelings)
ReadiLink™ xtra Rapid iFluor® 488 Antibody Labeling Kit *BSA-Compatible* (2 Labelings)
Catalog # AATB1955
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
ReadiLink™ xtra Rapid iFluor® 488 Antibody Labeling Kit *BSA-Compatible* (2 Labelings)
Catalog # AATB1955
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    2 Tests
  • Description:
    ReadiLink™ xtra rapid iFluor™ 488 antibody labelling kit 'BSA-compatible'

Specifications

About this item

ReadiLink™ xtra rapid antibody labelling kits require essentially only 2 simple mixing steps without a column purification needed.

  • Contains all essential components
  • Minimal hands on time
  • Ready to use
  • Robust and simple antibody labeling protocol
  • No column purification required
Caution: For research use only.