ContactUsLinkComponent
Cell Navigator® TMR Ceramide Golgi Staining Kit *Red Fluorescence*
Cell Navigator® TMR Ceramide Golgi Staining Kit *Red Fluorescence*
Catalog # AATB22752
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Cell Navigator® TMR Ceramide Golgi Staining Kit *Red Fluorescence*
Catalog # AATB22752
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Description:
    Cell Navigator™ TMR ceramide golgi staining kit *red fluorescence*
Specifications
About this item
The golgi apparatus is a complex of vesicles and folded membranes within the cytoplasm of most eukaryotic cells, involved in secretion and intracellular transport.
  • Live cell labeling
  • Improved signal
  • Multiple research applications
Caution: For research use only.