ReadiLink™ xtra Rapid XFD488 Antibody Labelling Kit *BSA-Compatible, XFD488 Same Structure to Alexa Fluor™ 488* (2 Labelings)
Catalog # AATB1978
Anbieter: AAT BIOQUEST
CAS-Nummer:
Technische Daten
- Pk:2 Tests
- Description:ReadiLink™ xtra rapid XFD488 antibody labelling kit 'BSA-compatible, XFD488 same structure to Alexa Fluor™ 488'
Technische Daten
Über diesen Artikel
XFD488 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 488 (Alexa Fluor® is the trademark of ThermoFisher).
- Contains all essential components
- Minimal hands on time
- Ready to use
- Robust and simple antibody labeling protocol
- No column purification required
Caution: For research use only.