ReadiLink™ Cy3 Oligo and ssDNA Labelling Kit
Supplier: AAT BIOQUEST
Certificates
About this item
The kit is optimised for efficient labelling and contains all the essential reagents required for efficient labelling of ssDNA or oligos.
- Robust protocol
- Bright labeling
The resulting Cy3-labelled DNA probes are ideally suited for biological applications, e.g., electrophoretic mobility shift assays (EMSA), Northern and Southern blots, colony or in situ hybridisations..
Packaging: Packaged in plastic vials.
Caution: For research use only.