ContactUsLinkComponent
Helixyte™ Green Fluorimetric Total Nucleic Acid Quantitation Kit
Helixyte™ Green Fluorimetric Total Nucleic Acid Quantitation Kit
Catalog # AATB17630
Supplier:  AAT Bioquest
CAS Number:  
undefined
Helixyte™ Green Fluorimetric Total Nucleic Acid Quantitation Kit
Catalog # AATB17630
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    200 Tests
  • Description:
    Kit is designed to measure total amounts of nucleic acids, including double-stranded DNA (dsDNA), single-stranded DNA (ssDNA) and RNA in an easy and accurate format
Specifications
About this item
Helixyte™ Green fluorimetric total nucleic acid quantitation kit is optimised for measuring the total amounts of nucleic acids with a fluorescence microplate reader.
  • Optimised for microplate readers
  • Sensitive and indiscriminate binding
The kit has all the essential reagents, including Helixyte™ Green all reagent, dilution buffer, and pre-diluted DNA standards..
Packaging: Plastic vial.
Caution: For research use only.