Order Entry
Switzerland
ContactUsLinkComponent
Cell Meter™ Live Cell TUNEL Apoptosis Assay Kit *Green Fluorescence* (50 Tests)
Cell Meter™ Live Cell TUNEL Apoptosis Assay Kit *Green Fluorescence* (50 Tests)
Catalog # AATB22849
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Meter™ Live Cell TUNEL Apoptosis Assay Kit *Green Fluorescence* (50 Tests)
Catalog # AATB22849
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    50 Tests
  • Description:
    Cell Meter™ live cell TUNEL apoptosis assay kit *green fluorescence*

Specifications

About this item

DNA fragmentation represents a characteristic of late stage apoptosis.

  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.