


Toluene : 2-Propanol : Water (49,5 : 50 : 0,5) % vol., VOLUSOL® titration solvent
Supplier: VWR Chemicals
Certificates
About this item
:
Flash Pt:10 °C (closed cup)
Storage Temperature:Ambient
UN:1993
ADR:3,II
:GHS02
:GHS08
:GHS07
Specification Test Results
- Assay (Toluene) (V/V):47.5 - 52.5 %
- Assay (Propan-2-ol) (V/V):47 - 52 %
- Water (V/V):0.4 - 0.6 %
Specification Test Results
Related Information
Certificates
Search by Lot Number2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 30L Metal drum for liquids UN X (ZPACK12124125) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951045)
Packaging Information
spec_VWRC5138.2500 spec_VWRC5138.2500 spec_VWRC5138.5000 spec_VWRC5138.9025 spec_VWRC5138.9025
Product specification
More
Frequently Bought Together