Order Entry
Switzerland
ContactUsLinkComponent
 
Milli-Q® CLX 7040 CLRW Water Purification System
Catalog # MILLZAFS51040
Supplier:  Merck
Milli-Q® CLX 7040 CLRW Water Purification System
Catalog # MILLZAFS51040
Supplier:  Merck

Specifications

  • Flow rate:
    Make-up flow rate 40 L/h
  • Description:
    Milli-Q® CLX 7040 CLRW water purification system
  • Environmentally Preferable:
  • Model:
    Milli-Q® CLX 7040
  • Feed water:
    Potable tap water
  • Production flow rate:
    4 L/min
  • Microorganisms:
    <1 cfu/ml
  • Power consumption:
    230 V / 50 - 60 Hz
  • Resistivity at +25 °C:
    >15 MΩ·cm
  • TOC content:
    <30 ppb
  • Weight:
    222 kg
  • W×D×H:
    543×797×1255 mm
  • Pk:
    1

Specifications

About this item

Milli-Q® CLX 7000 analyser feed systems produce up to 3000 L/day of pure water from tap.

  • Consistent CLRW (Clinical laboratory reagent water) quality water that exceeds CLSI® standards
  • Advanced and sustainable water purification technologies, including the Elix® EDI module and water-saving E.R.A.® technology
  • User-friendly touchscreen
  • Quiet, compact and mobile
  • Digital services for remote monitoring and traceability
  • Store in dry location

Milli-Q® Connect remote services simplifies system management, supports full data traceability and access, and facilitates audit preparation and lab accreditation.

Water filtered through 0,22 μm sterilising-grade membranes and exposed to bactericidal UV lamp.

Filter

Product Information