Order Entry
Switzerland
ContactUsLinkComponent
Proteinase K
Proteinase K
Supplier:  EUROCLONE
Certificates
Proteinase K
Supplier:  EUROCLONE

Specifications

  • CAS No.:
    39450-01-6
  • Storage conditions:
    Freezer
  • Enzyme name:
    Proteinase K

Specifications

Frequently Bought Together