Orders LinkContactUsLinkComponent
Q4ever™ Green
Q4ever™ Green
Supplier:   AAT Bioquest
Q4ever™ Green
Supplier:  AAT Bioquest
Certificates
Q4ever™ Green
2 Options Available Below
Product Details & Documents
A real-time polymerase chain reaction (real-time PCR), also known as quantitative polymerase chain reaction (qPCR), is a popular laboratory technique of molecular biology based on the polymerase chain reaction (PCR).
  • No probe required
  • Enhanced sensitivity
  • Low enzymatic inhibition
  • Binds to any dsDNA
Caution: For research use only. Store at −20 °C, protected from light. Product is stable for at least 12 months from the date of receipt when stored as recommended.
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • AATB17608
  • AATB17609
  • Application:
  • qPCR probe
  • qPCR probe
Specifications
Product Family Options
Product Information
  • Details
    Pk
    Availability
    Price
  • AATB17608
    Q4ever™ Green
    Q4ever™ Green
    1250X DMSO solution
    100 µl
    Log in for availability
    Log in for availability
    1 * 100 µl
    1 * 100 µl - SFr. 217.00
    SFr. 217.00
    1 * 100 µl
       SFr. 217.00
    Specifications:
    More
    Specifications
    Cat. No.
    AATB17608
    Application
    qPCR probe
  • AATB17609
    Q4ever™ Green
    Q4ever™ Green
    1250X DMSO solution
    2 mL
    Log in for availability
    Log in for availability
    1 * 2 mL
    1 * 2 mL - SFr. 2’870.00
    SFr. 2’870.00
    1 * 2 mL
       SFr. 2’870.00
    Specifications:
    More
    Specifications
    Cat. No.
    AATB17609
    Application
    qPCR probe