Order Entry
Switzerland
ContactUsLinkComponent
Native Horseradish Horseradish peroxidase (from Serum), Spectinomycin
Native Horseradish Horseradish peroxidase (from Serum), Spectinomycin
Catalog # AATB50037
Supplier:  AAT Bioquest
CAS Number:  
undefined
Native Horseradish Horseradish peroxidase (from Serum), Spectinomycin
Catalog # AATB50037
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    1 mg
  • Conjugation:
    Spectinomycin
  • Protein/peptide type:
    Native
  • Source:
    Serum
  • Species:
    Horseradish
  • Protein synonyms:
    HRP
  • Protein/peptide name:
    Horseradish peroxidase

Specifications

About this item

Spectinomycin-HRP conjugate is used for developing an ELISA assay for quantifying spectinomycin.

  • High immunogenicity
  • Useful for antibody development
  • ELISA assay building block
Caution: For research use only.