Order Entry
Switzerland
ContactUsLinkComponent
 
Native Megathura crenulata Keyhole limpet hemocyanin (from Serum), 3-Amino-2-oxazolidine (AOZ)
  AATB50005
 :  
undefined
Native Megathura crenulata Keyhole limpet hemocyanin (from Serum), 3-Amino-2-oxazolidine (AOZ)
  AATB50005
 :  

 

  • Pack type:
    Vial
  • Pk:
    1 mg
  • Conjugation:
    3-Amino-2-oxazolidine (AOZ)
  • Protein/peptide type:
    Native
  • Source:
    Serum
  • Species:
    Megathura crenulata
  • Protein synonyms:
    KLH
  • Protein/peptide name:
    Keyhole limpet hemocyanin

 

 

3-Amino-2-oxazolidine (AOZ)-KLH Conjugate is used for developing anti-3-Amino-2-oxazolidine (AOZ) antibodies that are typically used in an ELISA assay.

  • High immunogenicity
  • Useful for antibody development
  • ELISA assay building block
 : For research use only.