Wheat Germ Agglutinin, XFD594
Catalog # AATB25509
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Vial
- Pk:1 mg
- Conjugation:Alexa Fluor® 594
- Protein/peptide type:Native
- Source:Triticum aestivum
- Species:Triticum aestivum
- Protein synonyms:WGA,lectin
- Protein/peptide name:Wheat Germ Agglutinin
Specifications
About this item
XFD594 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 594 (Alexa Fluor® is the trademark of ThermoFisher).
- Well-cited conjugate
- High affinity and selectivity
- Multiple applications
Caution: For research use only.