Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), iFluor® 700
Supplier: AAT Bioquest
Certificates
About this item
AAT Bioquest's iFluor® dyes are optimised for labeling proteins, in particular, antibodies.
- Optimized for flow cytometry
- Useful for assay development
- High affinity
- Multiple applications
Caution: For research use only.
Specifications
- Conjugation:iFluor® 700
- Protein/peptide type:Native
- Source:Streptomyces avidinii
- Species:Streptomyces avidinii
- Protein synonyms:Avidin
- Protein/peptide name:Streptavidin
Specifications
Frequently Bought Together