Order Entry
Switzerland
ContactUsLinkComponent
Native Armoracia rusticana HRP (from Armoracia rusticana), Sulfacetamide
Native Armoracia rusticana HRP (from Armoracia rusticana), Sulfacetamide
Catalog # AATB50031
Supplier:  AAT Bioquest
CAS Number:  
undefined
Native Armoracia rusticana HRP (from Armoracia rusticana), Sulfacetamide
Catalog # AATB50031
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    1 mg
  • Conjugation:
    Sulfacetamide
  • Protein/peptide type:
    Native
  • Source:
    Armoracia rusticana
  • Species:
    Armoracia rusticana
  • Protein synonyms:
    Horseradish Peroxidase
  • Protein/peptide name:
    HRP

Specifications

About this item

Sulfacetamide-HRP conjugate is used for developing an ELISA assay for quantifying sulfacetamide.

  • Assay development
  • High sensitivity
Caution: For research use only.