Native Amanita phalloides Phalloidin (from Eukaryotic cells), iFluor™ 514
Catalog # AATB23116
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Vial
- Pk:300 Tests
- Conjugation:iFluor® 514
- Protein/peptide type:Native
- Source:Eukaryotic cells
- Species:Amanita phalloides
- Protein synonyms:Amanita phalloides
- Protein/peptide name:Phalloidin
Specifications
About this item
This yellow fluorescent phalloidin conjugate (equivalent to Alexa Fluor® 514-labeled phalloidin) selectively binds to F-actins.
- Suitable for all fluorescent imaging platforms
- Specific for actin in both living or fixed cells
- Multiple applications
Caution: For research use only.