Native N6-Methyladenosine (from Eukaryotic cells), Biotin
Catalog # AATB17100
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Vial
- Pk:1 mg
- Conjugation:Biotin
- Protein/peptide type:Native
- Source:Eukaryotic cells
- Protein synonyms:N6-Methyladenosine
- Protein/peptide name:N6-Methyladenosine
Specifications
About this item
N6-Methyladenosine is an abundant modification in mRNA and is found within some viruses, and most eukaryotes including mammals, insects, plants and yeast.
- Common RNA modification
- Multiple applications
Caution: For research use only.