Order Entry
Switzerland
ContactUsLinkComponent
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), iFluor® 790
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), iFluor® 790
  AATB16975
 :  
undefined
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), iFluor® 790
  AATB16975
 :  

 

  • Pack type:
    Vial
  • Pk:
    200 µG
  • Conjugation:
    iFluor® 790
  • Protein/peptide type:
    Native
  • Source:
    Streptomyces avidinii
  • Species:
    Streptomyces avidinii
  • Protein synonyms:
    Avidin|Streptomyces avidinii
  • Protein/peptide name:
    Streptavidin

 

 

AAT Bioquest's iFluor® dyes are optimised for labeling proteins, in particular, antibodies.

  • Extremely high biotin-binding affinity
  • High quality conjugate
 : For research use only.