Ac-DEVD-pNA
Catalog # AATB13405
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic Bottle
- Pk:5 mg
- Protein/peptide name:Ac-DEVD-pNA
- Purity:95%
Specifications
About this item
Ac-DEVD-pNA is a colourimetric substrate for caspase 3, a protease that is rapidly activated when cells are exposed to apoptotic conditions and that cleaves poly(ADP-ribose) polymerase.
- Hazard Class/ Packing Group: 5, II
- GHS symbol: GHS07
- Signal Word: Warning